• USA Home
  • P4152 - (2R,4R)-(−)-Pentanediol

EMAIL THIS PAGE TO A FRIEND
P4152 Sigma

(2R,4R)-(−)-Pentanediol

Purchase

Properties

storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
213.8 °F
Flash Point(C) 
101 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

243175 (R,R)-(−)-2,4-Pentanediol, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?