EMAIL THIS PAGE TO A FRIEND
P4160 Sigma

Proglumide

powder

Synonym: N-Benzoyl-N′,N′-dipropyl-DL-isoglutamine

  • CAS Number 6620-60-6

  • Empirical Formula (Hill Notation) C18H26N2O4

  • Molecular Weight 334.41

  •  EC Number 229-567-4

Purchase

Properties

form   powder

Description

Biochem/physiol Actions

CCK antagonist; selectively blocks CNS effects

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

M006 Proglumide sodium salt, solid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?