• USA Home
  • P4188 - Immobilon®-P Polyvinylidene difluoride membranes

EMAIL THIS PAGE TO A FRIEND
P4188 Sigma

Immobilon®-P Polyvinylidene difluoride membranes

L × W 10 cm × 10 cm

Synonym: PVDF

Purchase

Description

Application

Polyvinylidene difluoride membranes are popular membranes for immunoblotting (Western blotting), and sequencing of proteins. The basis of protein binding to PVDF is believed to be due to hydrophobic interactions. PVDF membranes for Western blotting are available in the most commonly used 0.45 μm pore size. PVDF membranes have a greater protein binding capacity, are more durable and more solvent resistant than nitrocellulose.

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?