• USA Home
  • P4233 - Hamilton® Softgrip® fixed-volume pipette
EMAIL THIS PAGE TO A FRIEND
P4233 Sigma
Hamilton® Softgrip® fixed-volume pipette
volume 200 μL
Purchase
Properties
Related Categories Hamilton, Hamilton Pipettors, Hamilton Softgrip Pipettes and tips, Labware, Liquid Handling Equipment,
description   At 100% volume (Accuracy within ±0.40%, Precision within ±0.18%)
feature   electronic: no
mfr. no.   Hamilton
volume   200 μL
color   mustard
Description
General description
Same ergonomic features as the Hamilton Softgrip adjustable volume pipette.
• Less discomfort in the hand and wrist as compared to other pipettes during prolonged use
• Multi-point color coding for easy volume identification
Every SoftGrip pipette is calibrated at Hamilton with standards traceable to NIST. A certificate of calibration documenting accuracy and precision is provided in the package along with non-wettable recalibration labels for field service and recalibration.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Softgrip is a registered trademark of Hamilton Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?