• USA Home
  • P4264 - 3-sn-Phosphatidylethanolamine from sheep brain

EMAIL THIS PAGE TO A FRIEND
P4264 Sigma

3-sn-Phosphatidylethanolamine from sheep brain

commercial grade, Type II-S

Synonym: 1,2-Diacyl-sn-glycero-3-phosphoethanolamine, L-α-Cephalin, L-α-Phosphatidylethanolamine from sheep brain

Purchase

Properties

Related Categories Biochemicals and Reagents, Glycerophosphoethanolamines, Glycerophospholipids, Lipids
grade   commercial grade
type   Type II-S
storage temp.   −20°C

Description

Biochem/physiol Actions

The major structural phospholipid in brain, comprising 20-25% of total lipid; primarily localized to gray matter.

Quality

A crude extract containing several phospholipids and glycolipids normally found in brain tissue.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

B3635 Brain Extract from bovine brain, Type VII

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?