• USA Home
  • P4328 - L-α-Phosphatidylcholine, β-arachidonoyl-γ-stearoyl

EMAIL THIS PAGE TO A FRIEND
P4328 Sigma

L-α-Phosphatidylcholine, β-arachidonoyl-γ-stearoyl

lyophilized powder

Purchase

Properties

form   lyophilized powder
shipped in   dry ice
storage temp.   −20°C

Description

Packaging

Packaged under Argon.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 1888 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

P1287 2-Arachidonoyl-1-stearoyl-sn-glycero-3-phosphocholine solution, 10 mg/mL in chloroform

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?