• USA Home
  • P4393 - 1,2-Dipalmitoyl-sn-glycerol 3-phosphate

EMAIL THIS PAGE TO A FRIEND
P4393 Sigma

1,2-Dipalmitoyl-sn-glycerol 3-phosphate

~99%

Synonym: 1,2-Dipalmitoyl-sn-glycero-3-phosphatidic acid, 3-sn-Phosphatidic acid, 1,2-dipalmitoyl, L-α-Phosphatidic acid, dipalmitoyl

  • CAS Number 7091-44-3

  • Empirical Formula (Hill Notation) C35H69O8P

  • Molecular Weight 648.89

  •  Beilstein Registry Number 1730519

Purchase

Properties

assay   ~99%
shipped in   dry ice
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

P4013 1,2-Dipalmitoyl-sn-glycero-3-phosphate sodium salt, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?