• USA Home
  • P4459 - Protein Kinase Cε Peptide

EMAIL THIS PAGE TO A FRIEND
P4459 Sigma

Protein Kinase Cε Peptide

lyophilized powder

Purchase

Description

Specificity

Synthetic peptide corresponding to the C-terminal variable (V5) region of rat PKC ε (726-737). Effective in competition experiments with Anti-Protein Kinase C ε (Sigma Product No. P 8458).

Physical form

Lyophilized from 0.1% TFA

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?