• USA Home
  • P4491 - Nunc® PCR amplification plates
EMAIL THIS PAGE TO A FRIEND
P4491 Sigma
Nunc® PCR amplification plates
96 well without skirt
Purchase
Description
General description
Nunc® PCR Amplification Plates
• Uniform thin wall construction of 0.2 mL well offers excellent heat transfer with maximum product yield for processing multiple PCR samples
• Rigid plate design without skirt fits GeNunc tube trade and holder (Nunc PCR amplificaiton workstation; product no. P4366), allowing easy manipulation by automated handling equipment
• Conical wells of plate extend above the base to permit sealing with strip caps or Amplification Tape 96 to prevent well to well contamination
• Strip caps have dome shape to eliminate condensation and use of oil overlays
• Amplification Tape 96 is offered in single, precut sheets
• Silicone Spacer Plate ensures even heat transfer when Amplification Tape 96 is used
• Plate and tape are certified RNAse and DNAse free
• Compatible with 96 well format thermal cyclers that hold a V-shaped aluminum block for 96 0.2 mL tubes
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?