• USA Home
  • P4529 - Pentaethylene glycol monohexyl ether

EMAIL THIS PAGE TO A FRIEND
P4529 Sigma

Pentaethylene glycol monohexyl ether

>90% (GC)

Synonym: C6E5, Hexyl pentaethylene glycol ether, Hexylpentaglycol, Polyoxyethylene (5) hexyl ether

  • CAS Number 86674-95-5

  • Linear Formula CH3(CH2)5(OCH2CH2)5OH

  • Molecular Weight 322.44

  •  Beilstein Registry Number 1911328

  •  MDL number MFCD00077944

  •  PubChem Substance ID 24898547

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents N to Z, Non-Ionic Detergents
description   non-ionic
InChI Key   SCRHZMGASXVJSJ-UHFFFAOYSA-N
assay   >90% (GC)

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?