• USA Home
  • P4534 - Poly(Orn, Tyr) 4:1 hydrobromide

EMAIL THIS PAGE TO A FRIEND
P4534 Sigma

Poly(Orn, Tyr) 4:1 hydrobromide

Orn:Tyr (4:1)

Purchase

Properties

feed ratio   Orn:Tyr (4:1)
mol wt   mol wt 20,000-50,000
storage temp.   −20°C

Description

Analysis Note

Molecular weights based on viscosity.7

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?