• USA Home
  • P4685 - IgG1−PE Isotype Control from murine myeloma

EMAIL THIS PAGE TO A FRIEND
P4685 Sigma

IgG1−PE Isotype Control from murine myeloma

clone MOPC 21, purified immunoglobulin, buffered aqueous solution

Synonym: IgG1 Isotype Control from murine myeloma

Purchase

Properties

Related Categories Analytical Cytology, Antibodies, Antibody Characterization and Analysis, Biochemicals and Reagents, Isotype Controls,
biological source   mouse
antibody form   purified immunoglobulin
clone   MOPC 21, monoclonal
form   buffered aqueous solution
application(s)   flow cytometry: suitable
isotype   IgG1κ
conjugate   phycoerythrin (R-PE) conjugate
shipped in   wet ice
storage temp.   2-8°C

Description

Preparation Note

Prepared by conjugation to R-phycoerythrin (PE). This orange-red dye is efficiently excited at 488 nm and emits at 578 nm.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Application

IgG1-PE Isotype Control antibodies are suitable for use in flow cytometry.

General description

Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases . Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice . IgG1-PE Isotype Control antibodies are specific for anti-mouse whole serum and anti-mouse IgG1. The product does not react with antiserum from mouse IgA, IgM, IgG2a, IgG2b, IgG3 and λ light chain.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?