• USA Home
  • P4709 - Protein Kinase Cδ Peptide

EMAIL THIS PAGE TO A FRIEND
P4709 Sigma

Protein Kinase Cδ Peptide

lyophilized powder

Purchase

Description

Other Notes

Lyophilized from 0.1% TFA in H2O

Specificity

Synthetic peptide, corresponding to the C-terminal variable (V5) region of rat PKC δ (662-673, with N-terminal added Lys). Effective in competition experiments with Anti-Protein Kinase C δ (Sigma Product No. P 8333).

Physical form

Lyophilized from 0.1% TFA

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?