• USA Home
  • P4725 - L-α-Phosphatidyl-D-myo-inositol 3,4-diphosphate, dioctanoyl

EMAIL THIS PAGE TO A FRIEND
P4725 Sigma

L-α-Phosphatidyl-D-myo-inositol 3,4-diphosphate, dioctanoyl

  • Empirical Formula (Hill Notation) C25H49O19P3

  • Molecular Weight 746.57

  •  MDL number MFCD02259149

  •  PubChem Substance ID 24898494

Purchase

Description

Biochem/physiol Actions

Activates Ca2+-insensitive PKC isotypes δ, ε, and η, activates Akt (a serine-threonine kinase also known as PKBα) by direct interaction with the Akt pleckstrin homology domain.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?