• USA Home
  • P4797 - Polyadenylic acid–Decathymidylic acid sodium salt

EMAIL THIS PAGE TO A FRIEND
P4797 Sigma

Polyadenylic acid–Decathymidylic acid sodium salt

powder

Synonym: Poly(A) • p(dT)10

Purchase

Description

Other Notes

Base ratio approx. 1:1

Unit Definition

One unit will yield an A260 of 1.0 in 1.0 ml of water (1 cm light path).

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?