• USA Home
  • P4806 - Protease from Bacillus sp.

EMAIL THIS PAGE TO A FRIEND
P4806 Sigma

Protease from Bacillus sp.

Purchase

Properties

storage temp.   −20°C

Description

Unit Definition

One unit will hydrolyze casein to produce color equivalent to 1.0 μmole (181 μg) of tyrosine per min at pH 7.5 at 37°C (color by Folin-Ciocalteu reagent), unless otherwise indicated.

Protocols & Applications

Non-specific Protease Activity Assay Video

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
UK9595000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Protocols

Enzymatic Assay of Protease Using Casein As a Substrate

To standardize a procedure for the enzymatic assay of Protease using Casein as a substrate at Sigma-Aldrich St. Louis.

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?