• USA Home
  • P4911 - Sigma adjustable volume digital single channel pipette
EMAIL THIS PAGE TO A FRIEND
P4911 Sigma
Sigma adjustable volume digital single channel pipette
capacity 10-100 μL , in 0.1 μl increments
Purchase
Properties
Related Categories Labware, Liquid Handling Equipment, Pipettes, Pipettors and Tips, Pipettors, Sigma Pipettors More...
availability   not available in EU
capacity   10-100 μL , in 0.1 μl increments
Description
General description
Volume change is accomplished by turning the plunger knob and reading the digital counter. A click-stop mechanism fixes the selected volume.
Each pipet has a built-in tip ejector.
Includes full specifications, operating instructions, spare O-rings, and a maintenance kit.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?