• USA Home
  • P4927 - Carestream® Kodak® black and white paper
EMAIL THIS PAGE TO A FRIEND
P4927 Sigma
Carestream® Kodak® black and white paper
Polycontrast III RC paper, sheet size 8 in. × 10 in.
Purchase
Description
General description
Fast, medium-weight enlarging paper with wide contrast range and smooth, glossy surface. Resin-coated base for rapid processing and drying. Process in Royalprint Model 417 or Dektomatic 65 Processors, or in trays with Dektol Developer. Safelight filter OC.
Legal Information
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Polycontrast is a trademark of Eastman Kodak Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?