• USA Home
  • P5026 - 20-Deoxy-20-oxophorbol 12,13-dibutyrate

EMAIL THIS PAGE TO A FRIEND
P5026 Sigma

20-Deoxy-20-oxophorbol 12,13-dibutyrate

Synonym: PDBuAL

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C13-C60, Carbonyl Compounds, Chemical Synthesis,
InChI Key   BSSVCYJMQUYSRI-YVQNUNKESA-N
storage temp.   −20°C

Description

Application

Reduction with tritiated NaBH4 yields tritiated phorbol 12,13-dibutyrate that can be used for receptor binding studies

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?