• USA Home
  • P5027 - Protease from Rhizopus sp.

EMAIL THIS PAGE TO A FRIEND
P5027 Sigma

Protease from Rhizopus sp.

Type XVIII, 0.2-0.6 unit/mg solid, pH 3.0, 37 °C

Synonym: Newlase

Purchase

Properties

type   Type XVIII
storage temp.   2-8°C

Description

Unit Definition

One unit will hydrolyze casein to produce color equivalent to 1.0 μmole (181 μg) of tyrosine per min at pH 7.5 at 37 °C (color by Folin-Ciocalteu reagent), unless otherwise indicated.

General description

Exhibits both proteolytic and lipolytic activities.

Stable in the acid range of pH 3-5. pH Optimum is 3.0.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
1
RTECS 
UK9540000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?