• USA Home
  • P5108 - Hamilton® Softgrip® adjustable volume pipette
EMAIL THIS PAGE TO A FRIEND
P5108 Sigma
Hamilton® Softgrip® adjustable volume pipette
2.5-25.0 μL volume, resolution: 0.02 μL
Purchase
Properties
Related Categories Hamilton, Hamilton Pipettors, Hamilton Softgrip Pipettes and tips, Labware, Liquid Handling Equipment,
feature   electronic: no
mfr. no.   Hamilton
measuring range   10% volume accuracy, accuracy: 4.50%, precision: 1.50%
  100% volume accuracy, accuracy: 0.80%, precision: 0.20%
  2.5-25.0 μL volume, resolution: 0.02 μL
volume   25 μL
color   forest green
Description
General description
SoftGrip pipettes set new standards for quality, precision, and comfort in manual pipetting. The soft, non-slip handle minimizes fatigue associated with repetitive, pipetting motions and its ergonomic shape is sized to perfectly fit your hand. The sloped plunger button is designed with a short stroke and low spring tension to reduce hand strain and motion.
The separate, easy-to-reach, tip ejector button is yet another convenient feature of these state-of-the-art pipettes. You′ll feel these proven differences:
• 50% less actuation force with a corresponding reduction in risk factors associated with musculoskeletal disorders, such as carpal tunnel syndrome
• Less user fatigue due to the soft ergonomic shape and lower plunger forces
• Less discomfort in the hand and wrist as compared to other pipettes during prolonged use
Every SoftGrip pipette is calibrated at Hamilton with standards traceable to NIST. A certificate of calibration documenting accuracy and precision is provided in the package along with non-wettable recalibration labels for field service and recalibration.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Softgrip is a registered trademark of Hamilton Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?