• USA Home
  • P5200 - Plex detection subkit for 60,000 cm2 of membrane

EMAIL THIS PAGE TO A FRIEND
P5200 Sigma

Plex detection subkit for 60,000 cm2 of membrane

Purchase

Properties

shipped in   wet ice
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?