• USA Home
  • P5203 - 2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphoethanolamine

EMAIL THIS PAGE TO A FRIEND
P5203 Sigma

2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphoethanolamine

lyophilized powder

Synonym: 1-Hexadecanoyl-2-(cis-9-octadecenoyl)-sn-glycero-3-phosphoethanolamine, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanolamine, 3-sn-Phosphatidylethanolamine, 2-Oleoyl-1-palmitoyl, L-β-Oleoyl-γ-palmitoyl-α-cephalin, POPE

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Glycerophospholipid Metabolism, Glycerophospholipids, Glycerophosphonoethanolamines,
InChI Key   FHQVHHIBKUMWTI-ISLYRVAYSA-N
form   lyophilized powder
storage temp.   −20°C

Description

Packaging

Packaged under Argon.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?