• USA Home
  • P5242 - Monoclonal Anti-Phospholipase A2, Secretory Group V antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
P5242 Sigma

Monoclonal Anti-Phospholipase A2, Secretory Group V antibody produced in mouse

clone MCL-3G1, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-HVPLA2

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Lipids in Cell Signaling, PH-PI,
species reactivity   human
application(s)   microarray: suitable
  western blot: 1-2 μg/mL using total cell extract from the mouse macrophage cell line P388
clone   MCL-3G1, monoclonal
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG1
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... PLA2G5(5322)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

recombinant W79A mutant of hVPLA2.

Physical form

Solution in phosphate buffered saline containing 1% BSA and 15 mM sodium azide.

Price and Availability


Detect Your Unseen Protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?