• USA Home
  • P5298 - Pro-Leu-Gly hydroxamate hydrochloride

EMAIL THIS PAGE TO A FRIEND
P5298 Sigma

Pro-Leu-Gly hydroxamate hydrochloride

  • CAS Number 120928-08-7

  • Empirical Formula (Hill Notation) C13H24N4O4 · HCl

  • Molecular Weight 336.82

  •  MDL number MFCD00238157

  •  PubChem Substance ID 24898617

Purchase

Description

Amino Acid Sequence

Pro-Leu-Gly-NHOH

Application

Affinity ligand for the purification of human collagenases.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?