• USA Home
  • P5329 - Protein Kinase Cβ2 Peptide

EMAIL THIS PAGE TO A FRIEND
P5329 Sigma

Protein Kinase Cβ2 Peptide

lyophilized powder

Synonym: Cys-Ser-Phe-Val-Asn-Ser-Glu-Phe-Leu-Lys-Pro-Glu-Val-Lys-Ser, PKC β2

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Enzyme Fragments, Peptides and Proteins, Peptides for Cell Biology,
InChI Key   URBJSHDJILNKFN-HKINMLJPSA-N
form   lyophilized powder
storage temp.   −20°C
Gene Information   human ... PRKCB(5579)

Description

Specificity

Synthetic peptide corresponding to the C-terminal region of PKC β2 (660-673).

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?