• USA Home
  • P5350 - N-CMV-30 Sequencing Primer

EMAIL THIS PAGE TO A FRIEND
P5350 Sigma

N-CMV-30 Sequencing Primer

Purchase

Properties

grade   for molecular biology
shipped in   dry ice
storage temp.   −20°C

Description

Components

The primer is supplied at an initial concentration of 5 uM in 1xTE buffer. The nucleotide sequence is as follows: 5′-AAT-GTC-GTA-ATA-ACC-CCG-CCC-CGT-TGA-CGC-3′

Application

The N-CMV-30 sequencing primer is suitable for sequencing from the C-terminus of pFLAG-CMV expression vectors.

General description

The N-CMV-30 sequencing primer is a chemically synthesized, 30 base, single strand oligodeoxyribonucleotide designed for dsDNA or ssDNA sequencing of FLAG® fusion proteins at the C-terminus of the pFLAG-CMV expression vectors. The primer is complementary to map position nucleotides 818-847 of positive, coding DNA strand of the pFLAG-CMV vectors.

Other Notes

(5′-AAT GTC GTA ATA ACC CCG CCC CGT TGA CGC-3′)

Packaging

1 μg in poly bottle

Legal Information

FLAG is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
HG1933000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

P5475 C-CMV-24 Sequencing Primer
P7957 C-24 Sequencing Primer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?