• USA Home
  • P5391 - Palmitoylcholine iodide

EMAIL THIS PAGE TO A FRIEND
P5391 Sigma

Palmitoylcholine iodide

  • CAS Number 26640-69-7

  • Linear Formula C21H44NO2I

  • Molecular Weight 469.48

  •  PubChem Substance ID 329820228

Purchase

Properties

InChI Key   PGZMGRIWQPBVCB-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?