EMAIL THIS PAGE TO A FRIEND
P5406 Sigma

S-Propylglutathione

  • CAS Number 24425-53-4

  • Empirical Formula (Hill Notation) C13H23N3O6S

  • Molecular Weight 349.40

  •  MDL number MFCD00056732

  •  PubChem Substance ID 24898636

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides and Tripeptides, Glutathione and Derivatives, Peptides
InChI Key   IJQSYVSEZCHJFD-UHFFFAOYSA-N
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?