EMAIL THIS PAGE TO A FRIEND
P5432 Sigma
Pipette holder
Hood mounted
Purchase
Properties
Related Categories Labware, Liquid Handling Equipment, Pipettes, Pipettes, Pipettors and Tips, Serological Pipettes More...
W × D × H   4 in. × 10 in. × 23 in.
Description
General description
Five compartment holder supplied with industrial strength, double-sided tape. Easily mounted on either the left or right side of hood.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z676039 Manual pipette racks, blue magnetic rack
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?