• USA Home
  • P5439 - Sartorius Proline® multichannel electronic pipettor
EMAIL THIS PAGE TO A FRIEND
P5439 Sigma
Sartorius Proline® multichannel electronic pipettor
volume 5-100 μL, 8 channel with Euro plug
Purchase
Properties
Related Categories BioHit Pipettors, Labware, Liquid Handling Equipment, Pipettes, Pipettors and Tips, Pipettors More...
AC input   230 V
mfr. no.   Sartorius, 710211
volume   5-100 μL
Description
General description
Sartorius Proline Electronic Multi-Channel pipettors of the Biohitfamily offer a wide volume range while delivering high accuracy and precision.
In addition the Proline Electronic Multi-Channels include individual piston/tip cone assemblies allowing easy repair and maintenance and a dispensing head that rotates clockwise or counter-clockwise for optimum Pipetting convenience.
Sartorius Proline electronic pipettors offer effortless and rapid pipetting with high levels of accuracy and precision. They offer a wide variety of pipetting operations, making the Proline range the complete solution for all pipetting needs. Proline pipettors are ergonomically designed and the tip ejection is easily operated with four fingers to avoid strain and fatigue during pipetting. A microprocessor-controlled piston ensures optimal precision and accuracy at the same time being user-independent, delivering reliable results consistently. Proline liquid handling operations include:
• Forward Pipetting
• Reverse Pipetting (excl. 1200)
• Multiple-Dispensing
• Multi-Aspirating (only 1200)
• Diluting (excl. mcp 1200)
• Sequential-Dispensing (only 1200)
• Post-Delivery Mixing
Legal Information
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?