EMAIL THIS PAGE TO A FRIEND
P5523 Sigma

Phe-Leu-Glu-Glu-Leu

≥97% (HPLC)

  • CAS Number 69729-06-2

  • Empirical Formula (Hill Notation) C31H47N5O10

  • Molecular Weight 649.73

  •  MDL number MFCD00076877

  •  PubChem Substance ID 24898663

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
InChI Key   JNBCIUTTWUUDDX-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Packaging

100 mg in glass bottle

Substrates

Substrate for vitamin K-dependent carboxylation studies.

Price and Availability


Detect Your Unseen Protein

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?