• USA Home
  • P5563 - Monoclonal Anti-Rat Lambda Light Chains−R-Phycoerythrin antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
P5563 Sigma

Monoclonal Anti-Rat Lambda Light Chains−R-Phycoerythrin antibody produced in mouse

clone RL-6, purified immunoglobulin, buffered aqueous solution

Synonym: Monoclonal Anti-Rat Lambda Light Chains

Purchase

Properties

Related Categories Alphabetical Index, Analytical Cytology, Anti-Immunoglobulins, Antibodies, Phycoerythrin (R-PE),
biological source   mouse
antibody form   purified immunoglobulin
clone   RL-6, monoclonal
form   buffered aqueous solution
storage condition   protect from light
application(s)   flow cytometry: 10 μL using 1 × 106 cells
isotype   IgG2a
conjugate   phycoerythrin (R-PE) conjugate
shipped in   wet ice
storage temp.   2-8°C

Description

Specificity

The antibody reacts with an epitope on the rat λ light chain found in various rat immunoglobulin classes and subclsasses. No cross-reactivity is observed with rat κ chains or with IgG from human, mouse, bovine, rabbit, goat, sheep, horse, dog, chicken, pig or cat. Weak cross-reactivity is observed with guinea pig IgG.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

How to Choose a Secondary Antibody

The following information is provided to help you decide which secondary antibody may be best for your particular application.
Keywords: Amplification, Enzyme-linked immunosorbent assay, Flow cytometry, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western blot

Secondary Antibodies, Conjugates and Kits

Secondary antibodies are polyclonal or monoclonal antibodies that bind to primary antibodies or antibody fragments, such as the Fc or Fab regions. They are typically labeled with probes that make the...
Keywords: Absorption, Adsorption, Amplification, Bacterial conjugations, Cancer, Digestions, Electrophoresis, Enzyme-linked immunosorbent assay, Flow cytometry, Gel electrophoresis, Gene expression, Hormones, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunology, Immunoprecipitation, Infrared spectroscopy, Microbiology, Microscopy, Purification, Vitamins, Western blot

Related Content

Antibody Explorer - Over 80,000 Products

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?