• USA Home
  • P5579 - Protein Kinase Cγ Peptide

EMAIL THIS PAGE TO A FRIEND
P5579 Sigma

Protein Kinase Cγ Peptide

lyophilized powder

Synonym: Cys[ACM]-Asp-Ala-Arg-Ser-Pro-Thr-Ser-Pro-Val-Pro-Val-Pro-Val-Met, PKC γ

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Enzyme Fragments, Peptides and Proteins, Peptides for Cell Biology,
InChI Key   QWNLMQOHNMILPB-UHFFFAOYSA-N
form   lyophilized powder
storage temp.   −20°C

Description

Specificity

Synthetic peptide corresponding to the C-terminal region of PKC γ (684-697).

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?