• USA Home
  • P5583 - Podophyllin from Podophyllum peltatum

EMAIL THIS PAGE TO A FRIEND
P5583 Sigma

Podophyllin from Podophyllum peltatum

powder

Synonym: Podophyllum resin

Purchase

Properties

form   powder
color   brown
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
TP8925000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?