• USA Home
  • P5616 - Nunc® 384 well polystyrene plates
EMAIL THIS PAGE TO A FRIEND
P5616 Sigma
Nunc® 384 well polystyrene plates
white, non-sterile
Purchase
Properties
Related Categories 384 Well Plates, Cell Culture, Drug Discovery, Labware, Manufacturer Browser,
material   white
description   non-treated
sterility   non-sterile
mfr. no.   Nunc 264572
plate L × W   128 mm × 86 mm
Description
General description
Nunc® 384 Well Plates, Polystyrene
• Cell culture surface available (Nunclon® Δ)
• Non-treated surface for storage, assays and screening
• MaxiSorp and PolySorp® for immunoassays; ELISA
• Clear plates for colorimetric assays and sample storage
• White plates for luminescence and fluorescence assays
• Black plates for fluorescence assays
• White and black plates minimize crosstalk
• Rounded square wells eliminate wicking (capillary action)
• Flat bottom for optical reading
• Optimized for scintillation counting
• Working volume range: 10-100 μl/well
Legal Information
MaxiSorp is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
PolySorp is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
All labs need water
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
Z723002 Nunc® 384 well polystyrene plates, 384 well plate, non-treated, polystyrene, 120ul/well, pinchbar design, 100/cs
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?