• USA Home
  • P5713 - L-α-Phosphatidyl-D-myo-inositol 3,5-diphosphate, dipalmitoyl

EMAIL THIS PAGE TO A FRIEND
P5713 Sigma

L-α-Phosphatidyl-D-myo-inositol 3,5-diphosphate, dipalmitoyl

  • Empirical Formula (Hill Notation) C41H81O19P3

  • Molecular Weight 970.99

  •  MDL number MFCD02179209

  •  PubChem Substance ID 24898696

Purchase

Description

Biochem/physiol Actions

Component of lipid signaling pathway.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?