• USA Home
  • P5738 - Principles of Tissue Engineering, 2nd ed.

EMAIL THIS PAGE TO A FRIEND
P5738 Sigma

Principles of Tissue Engineering, 2nd ed.

  •  ISBN-10 0-12-436630-9

  •  ISBN-13 978-0-12-436630-5

Purchase

Properties

Related Categories Books and Software, Cell Culture Books, Labware More...
publication info   R. Lanza, ed., Academic Press, 2000, 995 pp., hard cover
mfr. no.   Elsevier

Description

General description

This edition combines prerequisites for a understanding of tissue growth and development, tools and theoretical information needed to design tissues and organs, and applications of tissue engineering to diseases affecting specific organ systems. Few experts would have predicted the explosion in knowledge of gene expression, cell growth and differentiation, variety of stem cells, new polymers and materials available, or introduction of the first tissue-engineered products into the marketplace. This provides detailed analysis of research on all of the major systems of the human body and has new chapters on fetal tissue engineering and the universal cell.

Table of Contents

Introduction to Tissue Engineering:
1. The History and Scope of Tissue Engineering.,
2. The Challenge of Imitating Nature.,
Part I: The Basis of Growth and Differentiation:
3. Organization of Cells into Higher Ordered Structures.,
4. Dynamics of Cell-ECM Interactions.,
5. Matrix Molecules and Their Ligands.,
6. Inductive Phenomena.,
7. Morphogenesis and Tissue Engineering.,
8. Cell Determination and Differentiation.,
Part II: In Vitro Control of Tissue Development:
9. Mechanical and Chemical Determinants of Tissue Development.,
10. Animal Cell Culture.,
11. Regulation of Cell Behavior by Matricellular Proteins.,
12. Growth Factors.,
13. Tissue Engineering Bioreactors.,
14. Tissue Assembly in Microgravity.,
Part III: In Vivo Synthesis of Tissues and Organs:
15. In Vivo Synthesis of Tissues and Organs.,
Part IV: Models for Tissue Engineering:
16. Organotypic and Histiotypic Models of Engineered Tissues.,
17. Quantitative Aspects of Tissue Engineering: Basic Issues in Kinetics, Transport, and Mechanics.,
Part V: Biomaterials in Tissue Engineering:
18. Patterning of Cells and Their Environment.,
19. Cell Interactions with Polymers.,
20. Matrix Effects., l
21. Polymer Scaffold Processing.,
22. Biodegradable Polymers.,
Part VI: Transplantation of Engineered Cells and Tissues:
23. Approaches to Transplanting Engineered Cells and Tissues.,
24. Cryopreservation.,
25. Immunomodulation.,
26. Immunoisolation.,
27. Engineering Challenges in Immunoisolation Device Development.,
Part VII: Fetal Tissue Engineering:
28. Fetal Tissue Engineering.,
29. Pluripotent Stem Cells.,
Part VIII: Gene Therapy:
30. Gene-Based Therapeutics.,
Part IX: Breast:
31. Breast Reconstruction.,
Part X: Cardiovascular System:
32. Blood Vessels.,
33. Small-Diameter Vascular Grafts.,
34. Cardiac Prostheses.,
Part XI: Cornea:
35. Cornea.,
Part XII: Endocrinology and Metabolism:
36. Bioartificial Pancreas.,
37. Parathyroid.,
Part XIII: Gastrointestinal System:
38. Alimentary Tract.
39. Liver.,
40. HepatAssist Liver Support System.,
41. Lineage Biology and Liver.,
Part XIV: Hematopoietic System:
42. Red Blood Cell Substitutes.
43. Lymphoid Cells.,
44. Hematopoietic Stem Cells.,
Part XV: Kidney and Genitourinary System:
45. Renal Replacement Devices.,
46. Genitourinary System.,
Part XVI: Musculoskeletal System:
47. Structural Tissue Engineering.,
48. Bone Regeneration through Cellular Engineering.,
49. Articular Cartilage Injury.
50. Tendons and Ligaments.,
51. Mechanosensory Mechanisms in Bone.,
52. Myoblast Therapy.,
Part XVII: Nervous System:
53. Protection and Repair of Hearing.,
54. Vision Enhancement Systems.
55. Brain Implants.,
56. Nerve Regeneration.,
57. Transplantation Strategies for Treatment of Spinal Cord Dysfunction and Injury.,
58. Neural Stem Cells.,
Part XVIII: Periodontal and Dental Applications:
59. Periodontal Applications.,
60. Regeneration of Dentin.,
Part XIX: Skin:
61. Wound Repair: Basic Biology to Tissue Engineering.,
62. Skin.,
63. Dermal Equivalents.,
Part XX: Womb:
64. Artificial Womb.,
Part XXI: Regulatory Issues:
65. Regulatory Considerations.,
Epilogue.
Index.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?