• USA Home
  • P5741 - Nunc® 384 well polystyrene plates

EMAIL THIS PAGE TO A FRIEND
P5741 Sigma

Nunc® 384 well polystyrene plates

384 well plate, non-treated, polystyrene, 120ul/well, pinchbar design, 30/cs

Synonym: 384 multiwell plate, 384 well microplate, 384 well microtiter plate, 384 well plate

Purchase

Properties

Related Categories 384 Well Plates, Cell Culture, Drug Discovery, Labware, Manufacturer Browser,
material   black polystyrene plate
  clear polystyrene wells flat bottom
sterility   non-sterile
feature   lid: No
packaging   pack of 10
  case of 30
mfr. no.   Nunc 262260
maximum volume   120 μL
plate L × W   128 mm × 86 mm
size   384 wells
working volume   100 μL
matrix active group   non-treated surface treatment

Description

General description

Nunc® 384 Well Plates, Polystyrene

• Cell culture surface available (Nunclon® Δ)
• Non-treated surface for storage, assays and screening
• MaxiSorp and PolySorp® for immunoassays; ELISA
• Clear plates for colorimetric assays and sample storage
• White plates for luminescence and fluorescence assays
• Black plates for fluorescence assays
• White and black plates minimize crosstalk
• Rounded square wells eliminate wicking (capillary action)
• Flat bottom for optical reading
• Optimized for scintillation counting
• Working volume range: 10-100 μl/well

Legal Information

MaxiSorp is a trademark of Thermo Fisher Scientific or its subsidiaries

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries

PolySorp is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?