• USA Home
  • P5763 - L-α-Phosphatidylcholine solution

EMAIL THIS PAGE TO A FRIEND
P5763 Sigma

L-α-Phosphatidylcholine solution

Type V-E, 100 mg/mL in chloroform/methanol (9:1)

Purchase

Properties

type   Type V-E
concentration   100 mg/mL in chloroform/methanol (9:1)
storage temp.   −20°C

Description

Linkage

The material is the same as the lyophilized powder, P 7318

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
OG7565000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

P2772 L-α-Phosphatidylcholine, from egg yolk, Type XI-E, 100 mg/mL in chloroform, ≥99%, solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?