EMAIL THIS PAGE TO A FRIEND
P5765 Sigma

Prostaglandin F

synthetic

Synonym: (9α,11α,13E,15S)-9,11,15-Trihydroxyprost-13-en-1-oic acid, PGF

  • CAS Number 745-62-0

  • Empirical Formula (Hill Notation) C20H36O5

  • Molecular Weight 356.50

  •  Beilstein Registry Number 2704846

  •  MDL number MFCD00077862

  •  PubChem Substance ID 24898704

Purchase

Description

Biochem/physiol Actions

Stable metabolite of prostacyclin

Price and Availability


The Future of Bioimaging

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
GY4569700
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?