• USA Home
  • P5787 - Phthalic acid diethyl ester

EMAIL THIS PAGE TO A FRIEND
P5787 Sigma

Phthalic acid diethyl ester

99%

Purchase

Properties

assay   99%
density   1.12 g/mL at 25 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
2
RTECS 
TI1050000
Flash Point(F) 
312.8 °F
Flash Point(C) 
156 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

524972 Diethyl phthalate, 99.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?