• USA Home
  • P5788 - Pyruvate kinase from chicken muscle

EMAIL THIS PAGE TO A FRIEND
P5788 Sigma

Pyruvate kinase from chicken muscle

Prestained

Purchase

Properties

Related Categories Biochemicals and Reagents, Protein Extracts, Proteins and Derivatives More...
mol wt   unstained protein mol wt 58,000 Da
storage temp.   2-8°C

Price and Availability


Your Key to Success
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?