• USA Home
  • P5800 - Pyrilamine maleate salt

EMAIL THIS PAGE TO A FRIEND
P5800 Sigma

Pyrilamine maleate salt

Synonym: Mepyramine maleate salt, N-(4-Methoxyphenyl)methyl-N′,N′-dimethyl-N-(2-pyridinyl)-1,2-ethanediamine maleate salt

  • CAS Number 59-33-6

  • Empirical Formula (Hill Notation) C17H23N3O · C4H4O4

  • Molecular Weight 401.46

  •  MDL number MFCD00069333

  •  PubChem Substance ID 329820235

Purchase

Properties

InChI Key   JXYWFNAQESKDNC-BTJKTKAUSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?