EMAIL THIS PAGE TO A FRIEND
P5865 Sigma

Rp-8-PIP-cAMPS

≥98% (HPLC), solid

Synonym: 8-Piperidinoadenosine-3′,5′-cyclic monophosphorothioate, Rp-isomer sodium salt

  • Empirical Formula (Hill Notation) C15H20N6O5PSNa

  • Molecular Weight 450.39

  •  MDL number MFCD04037244

  •  PubChem Substance ID 329820237

Purchase

Description

Biochem/physiol Actions

Site-selective, lipophilic analogue of the phosphodiesterase-stable protein kinase A inhibitor Rp-cAMPS (Prod. No. A-165).

Price and Availability


Better Together

Discover your Target

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?