• USA Home
  • P5910 - Phospholipase A2 from Crotalus durissus terrificus venom

EMAIL THIS PAGE TO A FRIEND
P5910 Sigma

Phospholipase A2 from Crotalus durissus terrificus venom

lyophilized powder, ~200 units/mg protein

Synonym: Lecithinase A, PLA2, Phosphatidylcholine 2-acylhydrolase

Purchase

Description

Analysis Note

Protein determined by biuret.

Biochem/physiol Actions

Hydrolyzes the β-ester bond of zwitterionic glycerophospholipids. Preferred substrates are phosphatidylcholine, phosphatidylethanolamine, and their plasmalogen analogues. Phosphatidylinositol and phosphatidylserine are also hydrolyzed. Aggressively attacks phospholipids in membranes of intact cells.

Unit Definition

One unit will hydrolyze 1.0 μmole of soybean L-α-phosphatidylcholine to L-α-lysophosphatidylcholine and a fatty acid per min at pH 8.9 at 25 °C.

Physical form

Lyophilized powder containing Tris buffer salts

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
SZ6114900
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

inhibitor

Product #

Description

Add to Cart

D8008 7,7-Dimethyl-(5Z,8Z)-eicosadienoic acid, ≥95%
D6518 Diethylenetriaminepentaacetic acid, ≥99% (titration)
D6908 DL-erythro-Dihydrosphingosine, ≥99%, synthetic
P9375 1,10-Phenanthroline monohydrate, reagent grade
E4378 Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
ED2SS Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?