EMAIL THIS PAGE TO A FRIEND
P5927 Sigma

TWEEN® 20

for electrophoresis, suitable for solubilizing agent of membrane proteins and as a blocking reagent in immunoblotting

Synonym: Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate

Purchase

Properties

Related Categories Antigen-Vaccine Preparation, Biochemicals and Reagents, Cell Culture, Cell Lysis and Protein Extraction Reagents, Core Bioreagents,
grade   for electrophoresis
description   non-ionic
form   liquid
mol wt   mol wt ~1228
composition   lauric acid, ≥40% (balance primarily myristic, palmitic, and stearic acids)
refractive index   n20/D 1.468(lit.)
CMC   60 mg/L
density   1.095 g/mL at 25 °C(lit.)
suitability   suitable for solubilizing agent of membrane proteins and as a blocking reagent in immunoblotting

Description

Application

Non-ionic detergent

Legal Information

TWEEN is a registered trademark of Croda International PLC

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Detect Your Unseen Protein
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
TR7400000
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?