• USA Home
  • P6022 - Protein Kinase A Regulatory subunit from bovine heart

EMAIL THIS PAGE TO A FRIEND
P6022 Sigma

Protein Kinase A Regulatory subunit from bovine heart

secondary activity: 50-100 (units per μg protein), lyophilized powder

Purchase

Properties

Related Categories Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes, Cell Signaling and Neuroscience,
form   lyophilized powder
mol wt   dimer mol wt 400 residues (each chain)
capacity   5-10 nmol/mg, protein binding capacity (cyclic-AMP)(of cyclic AMP bound at pH 7.0, 0 °C at 64 nM [cyclic-AMP].)
storage temp.   −20°C

Description

Packaging

Package size based on inhibitory units.

Quality

Residual phosphorylating activity of approx. 0.08 unit per μg protein is found at pH 6.5 at 30 °C.

Reconstitution

For full activity, reconstitute in water with 6 mg/ml DTT.

Unit Definition

One unit will inhibit one unit of "catalytic subunit" phosphorylating activity in the absence of cyclic-AMP.

Physical form

Lyophilized powder containing ≤ 1% protein with sucrose and phosphate buffer salts as stabilizer

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?