• USA Home
  • P6034 - 1-O-Palmityl-2-O-methyl-rac-glycero-3-phosphocholine

EMAIL THIS PAGE TO A FRIEND
P6034 Sigma

1-O-Palmityl-2-O-methyl-rac-glycero-3-phosphocholine

Synonym: DL-α-Phosphatidylcholine, β-O-methyl-γ-O-hexadecyl

  • CAS Number 85405-05-6

  • Empirical Formula (Hill Notation) C25H54NO6P

  • Molecular Weight 495.67

  •  MDL number MFCD00063523

  •  PubChem Substance ID 24898729

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Ether Lipid Metabolism, Glycerophospholipids, Glycerophosphonocholines,
InChI Key   URQVDVCLCFNJMQ-UHFFFAOYSA-N
storage temp.   −20°C

Description

Other Notes

A biologically active analog of platelet activating factor (L-α-phosphatidylcholine, β-acetyl-γ-O-alkyl).

Price and Availability


The Future of Bioimaging

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?