• USA Home
  • P6156 - Phosphoglucomutase preparation from chicken muscle

EMAIL THIS PAGE TO A FRIEND
P6156 Sigma

Phosphoglucomutase preparation from chicken muscle

ammonium sulfate suspension, ~100 units/mg protein

Synonym: α-D-Glucose-1,6-bisphosphatase, α-D-Glucose-1-phosphate phosphotransferase

Purchase

Properties

form   ammonium sulfate suspension
storage temp.   2-8°C

Description

Analysis Note

Protein determined by biuret.

Unit Definition

One unit will convert 1.0 μmole of α-D-glucose 1-phosphate to α-D-glucose 6-phosphate per min at pH 7.4 at 30°C.

Physical form

Crystalline suspension in 3.2 M (NH4)2SO4, pH 6, containing 0.01% EDTA

Price and Availability

Safety & Documentation

Safety Information

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

P3397 Phosphoglucomutase from rabbit muscle, ammonium sulfate suspension, ≥100 units/mg protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?